Gene Transcripts

Gene Symbol
Acox3
Entrez Gene ID
83522
Gene Name
acyl-CoA oxidase 3, pristanoyl
Gene Aliases
Chromosome Location
Chr. 14: 80313202 - 80358130 on Build Rnor_6.0
UniGene ID
Rn.10546
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053339.1 NP_445791.1 4 462
XM_008770372.1 XP_008768594.1 4 3168
XM_008770373.1 XP_008768595.1 4 3211
XM_008770374.1 XP_008768596.1 3 435
GenBank mRNA X95188.1 - 4 462

View Full Product Details