Gene Transcripts

Gene Symbol
Lancl1
Entrez Gene ID
114515
Gene Name
LanC lantibiotic synthetase component C-like 1 (bacterial)
Gene Aliases
GPR69A, p40
Chromosome Location
Chr. 9: 74018192 - 74048098 on Build Rnor_6.0
UniGene ID
Rn.38972
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053723.1 NP_446175.1 5 731
GenBank mRNA AJ131111.1 - 5 731
BC085811.1 - 5 709 7115423

View Full Product Details