Gene Transcripts

Gene Symbol
Gabra4
Entrez Gene ID
140675
Gene Name
gamma-aminobutyric acid (GABA) A receptor, alpha 4
Gene Aliases
Chromosome Location
Chr. 14: 39154541 - 39230994 on Build Rnor_6.0
UniGene ID
Rn.81205
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080587.3 NP_542154.3 8 1130
XM_008770135.1 XP_008768357.1 7 1817
GenBank mRNA S55933.1 - 8 1130

View Full Product Details