Gene Transcripts

Gene Symbol
Mpst
Entrez Gene ID
192172
Gene Name
mercaptopyruvate sulfurtransferase
Gene Aliases
Mst
Chromosome Location
Chr. 7: 119626636 - 119631317 on Build Rnor_6.0
UniGene ID
Rn.32263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138843.1 NP_620198.1 1 634
GenBank mRNA BC086575.1 - 1 449
D50564.1 - 1 634

View Full Product Details