Gene Transcripts

Gene Symbol
Tpsg1
Entrez Gene ID
302990
Gene Name
tryptase gamma 1
Gene Aliases
Chromosome Location
Chr. 10: 14727180 - 14731086 on Build Rnor_6.0
UniGene ID
Rn.163109
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175593.2 NP_783183.1 3 269
GenBank mRNA AY196208.1 - 3 269

View Full Product Details