Gene Transcripts

Gene Symbol
Ambp
Entrez Gene ID
25377
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
Chromosome Location
Chr. 5: 78975690 - 78986020 on Build Rnor_6.0
UniGene ID
Rn.18721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012901.1 NP_037033.1 4 375
XM_008763759.1 XP_008761981.1 4 461
GenBank mRNA BC088166.1 - 4 360
S87544.1 - 4 375

View Full Product Details