Gene Transcripts

Gene Symbol
Hmbs
Entrez Gene ID
25709
Gene Name
hydroxymethylbilane synthase
Gene Aliases
PBG-D, PBGD, URO-S, hemC
Chromosome Location
Chr. 8: 48667278 - 48674670 on Build Rnor_6.0
UniGene ID
Rn.11080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013168.2 NP_037300.2 8 597
XM_006242896.1 XP_006242958.1 8 683
XM_006242897.1 XP_006242959.1 7 605
XM_008766148.1 XP_008764370.1 7 767
XM_008766149.1 XP_008764371.1 7 704
GenBank mRNA BC088162.1 - 8 597
FQ225972.1 - 2 82
FQ226062.1 - 8 520
FQ230962.1 - 2 82
FQ231091.1 - 8 581
FQ234870.1 - 6 753
X06827.1 - 8 472
Y12006.1 - 8 467

View Full Product Details