Gene Transcripts

Gene Symbol
Kcnj6
Entrez Gene ID
25743
Gene Name
potassium channel, inwardly rectifying subfamily J, member 6
Gene Aliases
Chromosome Location
Chr. 11: 35011015 - 35262362 on Build Rnor_6.0
UniGene ID
Rn.10185
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013192.2 NP_037324.1 3 2173
GenBank mRNA AB073753.1 - 3 2173

View Full Product Details