Gene Transcripts

Gene Symbol
Hpn
Entrez Gene ID
29135
Gene Name
hepsin
Gene Aliases
Chromosome Location
Chr. 1: 89534112 - 89549575 on Build Rnor_6.0
UniGene ID
Rn.11139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017112.1 NP_058808.1 4 311
GenBank mRNA BC072688.1 - 4 215
FQ219040.1 - 4 206
X70900.1 - 4 311

View Full Product Details