Gene Transcripts

Gene Symbol
Hspa13
Entrez Gene ID
29734
Gene Name
heat shock protein 70 family, member 13
Gene Aliases
Stch
Chromosome Location
Chr. 11: 14146515 - 14160697 on Build Rnor_6.0
UniGene ID
Rn.11050
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019271.2 NP_062144.2 3 422
GenBank mRNA AF006617.1 - 3 422
BC070954.1 - 3 422 7103134

View Full Product Details