Gene Transcripts
- Gene Symbol
- Htr1a
- Entrez Gene ID
- 24473
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled
- Gene Aliases
- 5HT1A, RAT5HT1A
- Chromosome Location
- Chr. 2: 36246628 - 36247896 on Build Rnor_6.0
- UniGene ID
- Rn.44486
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012585.1 |
NP_036717.1
|
1 |
723 |
|
View Full Product Details