Gene Transcripts
- Gene Symbol
- PADI4
- Entrez Gene ID
- 23569
- Gene Name
- peptidyl arginine deiminase 4
- Gene Aliases
- PAD, PAD4, PADI5, PDI4, PDI5
- Chromosome Location
- Chr. 1: 17308195 - 17364004 on Build GRCh38
- UniGene ID
- Hs.522969
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012387.2 |
NP_036519.2
|
2, 3 |
309 |
|
XM_011541150.1 |
XP_011539452.1
|
2, 3 |
307 |
|
XM_011541151.1 |
XP_011539453.1
|
2, 3 |
309 |
|
XM_011541153.1 |
XP_011539455.1
|
2, 3 |
309 |
|
XM_011541154.2 |
XP_011539456.1
|
2, 3 |
309 |
|
XM_011541155.1 |
XP_011539457.1
|
2, 3 |
309 |
|
XM_011541156.1 |
XP_011539458.1
|
2, 3 |
309 |
|
GenBank mRNA |
AB017919.1 |
- |
2, 3 |
307 |
|
AF229067.1 |
- |
2, 3 |
310 |
|
AK309637.1 |
- |
2, 3 |
301 |
|
AK314839.1 |
- |
2, 3 |
309 |
|
BC025718.1 |
- |
2, 3 |
293 |
5228777 |
View Full Product Details