Gene Transcripts

Gene Symbol
Syp
Entrez Gene ID
24804
Gene Name
synaptophysin
Gene Aliases
Syp1
Chromosome Location
Chr. X: 15698843 - 15707424 on Build Rnor_6.0
UniGene ID
Rn.11067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC099798.1 - 1381
X06177.1 - 1375
X06388.1 - 1408
X06655.1 - 1339

View Full Product Details