Gene Transcripts

Gene Symbol
Apoa4
Entrez Gene ID
25080
Gene Name
apolipoprotein A-IV
Gene Aliases
Apo-AIV, ApoA-IV, apoAIV
Chromosome Location
Chr. 8: 50536983 - 50539371 on Build Rnor_6.0
UniGene ID
Rn.15739
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012737.1 NP_036869.1 3 651
GenBank mRNA BC091159.1 - 3 626
FQ218620.1 - 3 652
M00002.1 - 3 651

View Full Product Details