Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013250.2 | NP_037382.2 | 3 | 665 | |
XM_006718311.2 | XP_006718374.1 | 3 | 705 | ||
GenBank mRNA | AF056618.1 | - | 3 | 665 | |
AK298365.1 | - | 3 | 625 | ||
AK302639.1 | - | 3 | 708 | ||
BC014538.1 | - | 1 | 167 | 3827937 |