Gene Transcripts

Gene Symbol
TAS2R42
Entrez Gene ID
353164
Gene Name
taste 2 receptor member 42
Gene Aliases
T2R24, T2R42, T2R55, TAS2R55
Chromosome Location
Chr. 12: 11185993 - 11186937 on Build GRCh38
UniGene ID
Hs.553716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181429.2 NP_852094.2 1 128
GenBank mRNA BC131742.1 - 1 128

View Full Product Details