Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019076.4 | NP_061949.3 | 1 | 374 | |
GenBank mRNA | AF030310.1 | - | 1 | 311 | |
AF462267.1 | - | 1 | 344 | ||
AF462268.1 | - | 1 | 374 | ||
AK313488.1 | - | 1 | 357 | ||
AY435143.1 | - | 1 | 311 |