Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300905.1 | NP_001287834.1 | 2 | 275 | |
NM_013449.3 | NP_038477.2 | 2 | 266 | ||
XM_011537797.2 | XP_011536099.1 | 2 | 180 | ||
XM_011537798.2 | XP_011536100.1 | 2 | 180 | ||
XM_011537799.2 | XP_011536101.1 | 2 | 180 | ||
XM_011537800.2 | XP_011536102.1 | 2 | 232 | ||
XM_011537801.2 | XP_011536103.1 | 2 | 259 | ||
XM_011537803.2 | XP_011536105.1 | 2 | 275 | ||
XM_011537804.2 | XP_011536106.1 | 4 | 358 | ||
XM_011537805.2 | XP_011536107.1 | 3 | 361 | ||
XM_011537806.2 | XP_011536108.1 | 2 | 139 | ||
XM_011537807.2 | XP_011536109.1 | 2 | 180 | ||
XM_017018740.1 | XP_016874229.1 | 3 | 406 | ||
XM_017018741.1 | XP_016874230.1 | 3 | 180 | ||
XM_017018742.1 | XP_016874231.1 | 3 | 406 | ||
GenBank mRNA | AB002312.2 | - | 1 | 55 | |
AK023842.1 | - | 2 | 155 | ||
AK303327.1 | - | 2 | 240 | ||
CD686304.1 | - | 2 | 266 | ||
CR749379.1 | - | 2 | 267 | ||
DB215236.1 | - | 2 | 275 |