Gene Transcripts
- Gene Symbol
- MS4A6A
- Entrez Gene ID
- 64231
- Gene Name
- membrane spanning 4-domains A6A
- Gene Aliases
- 4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
- Chromosome Location
- Chr. 11: 60171607 - 60184666 on Build GRCh38
- UniGene ID
- Hs.523702
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001247999.1 |
NP_001234928.1
|
5, 6 |
898 |
|
NM_022349.3 |
NP_071744.2
|
4, 5 |
709 |
|
NM_152851.2 |
NP_690590.1
|
4, 5 |
709 |
|
NM_152852.2 |
NP_690591.1
|
5, 6 |
834 |
|
XM_005274177.2 |
XP_005274234.1
|
5, 6 |
893 |
|
XM_006718658.2 |
XP_006718721.1
|
5, 6 |
768 |
|
XM_006718660.2 |
XP_006718723.1
|
4, 5 |
633 |
|
XM_006718661.2 |
XP_006718724.1
|
4, 5 |
633 |
|
XM_017018123.1 |
XP_016873612.1
|
3, 4 |
369 |
|
XM_017018124.1 |
XP_016873613.1
|
3, 4 |
369 |
|
GenBank mRNA |
AB013104.1 |
- |
4, 5 |
417 |
|
AF142409.1 |
- |
4, 5 |
437 |
|
AF212240.1 |
- |
4, 5 |
578 |
|
AF237908.1 |
- |
4, 5 |
579 |
|
AF261136.1 |
- |
4, 5 |
579 |
|
AF286866.1 |
- |
3, 4 |
357 |
|
AF350502.1 |
- |
4, 5 |
426 |
|
AF350503.1 |
- |
4, 5 |
426 |
|
AF354930.1 |
- |
4, 5 |
579 |
|
AK291870.1 |
- |
5, 6 |
834 |
|
AK300725.1 |
- |
4, 5 |
443 |
|
AK301387.1 |
- |
3, 4 |
269 |
|
BC022854.1 |
- |
4, 5 |
448 |
3934920 |
BC108728.1 |
- |
4, 5 |
482 |
6196213 |
DA917243.1 |
- |
4, 5 |
533 |
|
DA979138.1 |
- |
4, 5 |
480 |
|
View Full Product Details