Gene Transcripts

Gene Symbol
MKNK1
Entrez Gene ID
8569
Gene Name
MAP kinase interacting serine/threonine kinase 1
Gene Aliases
MNK1
Chromosome Location
Chr. 1: 46557407 - 46607140 on Build GRCh38
UniGene ID
Hs.371594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135553.2 NP_001129025.1 13 1383
NM_003684.5 NP_003675.2 14 1506
NM_198973.3 NP_945324.1 12 1297
NR_024174.2 11 1265
NR_024176.2 13 1410
XM_006711000.1 XP_006711063.1 13 1336
XM_006711001.2 XP_006711064.1 14 1586
XM_006711002.2 XP_006711065.1 13 6811
XM_006711003.1 XP_006711066.1 14 1397
XM_006711006.3 XP_006711069.1 12 4126
XM_006711007.1 XP_006711070.1 12 1228
XM_006711009.3 XP_006711072.1 12 6713
XM_006711013.1 XP_006711076.1 12 1261
XM_006711014.1 XP_006711077.1 12 1252
XM_006711015.1 XP_006711078.1 13 1323
XM_017002652.1 XP_016858141.1 13 7059
XM_017002653.1 XP_016858142.1 14 1684
XM_017002654.1 XP_016858143.1 14 4536
XM_017002655.1 XP_016858144.1 14 7157
XM_017002656.1 XP_016858145.1 14 6956
XM_017002657.1 XP_016858146.1 13 6858
XM_017002658.1 XP_016858147.1 12 7015
XM_017002659.1 XP_016858148.1 12 1311
XM_017002660.1 XP_016858149.1 12 6767
XM_017002661.1 XP_016858150.1 13 1532
XM_017002662.1 XP_016858151.1 13 1353
XM_017002663.1 XP_016858152.1 13 7113
XM_017002664.1 XP_016858153.1 12 6737
XM_017002665.1 XP_016858154.1 11 1184
XM_017002666.1 XP_016858155.1 12 4168
XM_017002667.1 XP_016858156.1 10 1744
GenBank mRNA AB000409.1 - 13 1326
AB451300.1 - 12 1139
AB451432.1 - 12 1139
AK096423.1 - 11 972
AK290456.1 - 12 1259
AK298839.1 - 11 1239
AY355461.1 - 11 1095
BC002755.2 - 14 1426 3629765
BX538193.1 - 11 1133

View Full Product Details