Gene Transcripts

Gene Symbol
OR10Z1
Entrez Gene ID
128368
Gene Name
olfactory receptor family 10 subfamily Z member 1
Gene Aliases
OR1-15
Chromosome Location
Chr. 1: 158606439 - 158607380 on Build GRCh38
UniGene ID
Hs.553590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004478.1 NP_001004478.1 1 275

View Full Product Details