Gene Transcripts
- Gene Symbol
- NDUFAF7
- Entrez Gene ID
- 55471
- Gene Name
- NADH:ubiquinone oxidoreductase complex assembly factor 7
- Gene Aliases
- C2orf56, MidA, PRO1853
- Chromosome Location
- Chr. 2: 37231631 - 37259052 on Build GRCh38
- UniGene ID
- Hs.433466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083946.1 |
NP_001077415.1
|
5, 6 |
570 |
|
NM_144736.4 |
NP_653337.1
|
7, 8 |
864 |
|
XM_005264410.1 |
XP_005264467.1
|
7, 8 |
844 |
|
XM_005264411.1 |
XP_005264468.1
|
6, 7 |
629 |
|
XM_011532956.1 |
XP_011531258.1
|
7, 8 |
800 |
|
GenBank mRNA |
AK299015.1 |
- |
6, 7 |
625 |
|
AK302150.1 |
- |
6, 7 |
734 |
|
AK303555.1 |
- |
6, 7 |
753 |
|
BC004548.2 |
- |
7, 8 |
829 |
3959139 |
BC012374.1 |
- |
2, 3 |
178 |
|
BI334629.1 |
- |
7, 8 |
658 |
|
BX538031.1 |
- |
5, 6 |
523 |
|
View Full Product Details