Gene Transcripts

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
Chromosome Location
Chr. 1: 231332731 - 231337872 on Build GRCh38
UniGene ID
Hs.356198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175876.4 NP_787072.2 1 1255
GenBank mRNA AK096460.1 - 1 1235
BC035763.1 - 1 1201 5552837

View Full Product Details