Gene Transcripts

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
Chromosome Location
Chr. 4: 119296419 - 119304445 on Build GRCh38
UniGene ID
Hs.173705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001701.3 NP_001001701.2 2 701
NM_001170330.1 NP_001163801.1 3 822
GenBank mRNA AF075061.1 - 2 509
AK025703.1 - 2 465
BC017399.1 - 2 524 4108498
CN361054.1 - 2 252

View Full Product Details