Gene Transcripts

Gene Symbol
Sdad1
Entrez Gene ID
289504
Gene Name
SDA1 domain containing 1
Gene Aliases
Chromosome Location
Chr. 14: 17247381 - 17274228 on Build Rnor_6.0
UniGene ID
Rn.12755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006958.1 NP_001006959.1 14, 15 1254
XM_006250722.2 XP_006250784.1 14, 15 1324
GenBank mRNA BC083620.1 - 14, 15 1254

View Full Product Details