Gene Transcripts

Gene Symbol
Matn1
Entrez Gene ID
297894
Gene Name
matrilin 1, cartilage matrix protein
Gene Aliases
Chromosome Location
Chr. 5: 149077412 - 149086957 on Build Rnor_6.0
UniGene ID
Rn.15159
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006979.1 NP_001006980.1 4 796
GenBank mRNA BC083869.1 - 4 796

View Full Product Details