Gene Transcripts

Gene Symbol
St7l
Entrez Gene ID
295344
Gene Name
suppression of tumorigenicity 7-like
Gene Aliases
Chromosome Location
Chr. 2: 207350651 - 207421082 on Build Rnor_6.0
UniGene ID
Rn.102337
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007639.1 NP_001007640.1 12, 13 2012
GenBank mRNA BC079194.1 - 12, 13 2012

View Full Product Details