Gene Transcripts

Gene Symbol
Fbxo30
Entrez Gene ID
308283
Gene Name
F-box protein 30
Gene Aliases
Chromosome Location
Chr. 1: 5366120 - 5389560 on Build Rnor_6.0
UniGene ID
Rn.204927
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007690.1 NP_001007691.1 2 1082
XM_006227640.2 XP_006227702.1 2 1124
GenBank mRNA BC083823.1 - 2 1082

View Full Product Details