Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008495.3 | NP_001008495.2 | 3 | 1002 | |
NM_001146273.1 | NP_001139745.1 | 2 | 846 | ||
GenBank mRNA | AK123958.1 | - | 3 | 696 | |
AL834364.1 | - | 3 | 708 | ||
AY147881.1 | - | 3 | 807 | ||
BC113828.1 | - | 6 | 732 |