Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008526.2 | NP_001008526.1 | 809 | ||
XM_006240601.2 | XP_006240663.1 | 969 | |||
XM_006240602.2 | XP_006240664.1 | 909 | |||
XM_006240603.2 | XP_006240665.1 | 885 | |||
GenBank mRNA | AY366364.1 | - | 854 | ||
BC088418.1 | - | 809 | 7123242 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008764977.1 | XP_008763199.1 | 1 | 698 | |
XM_008764978.1 | XP_008763200.1 | 2 | 867 | ||
XM_008764979.1 | XP_008763201.1 | 2 | 883 | ||
XM_008764980.1 | XP_008763202.1 | 2 | 885 |