Gene Transcripts

Gene Symbol
ZFYVE16
Entrez Gene ID
9765
Gene Name
zinc finger FYVE-type containing 16
Gene Aliases
PPP1R69
Chromosome Location
Chr. 5: 80407615 - 80479869 on Build GRCh38
UniGene ID
Hs.482660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105251.2 NP_001098721.1 4 611
NM_001284236.1 NP_001271165.1 4 537
NM_001284237.1 NP_001271166.1 3 483
NM_014733.4 NP_055548.3 3 483
XM_005248632.4 XP_005248689.1 3 609
XM_011543753.2 XP_011542055.1 4 508
XM_017010091.1 XP_016865580.1 5 669
XM_017010092.1 XP_016865581.1 4 655
XM_017010095.1 XP_016865584.1 4 508
GenBank mRNA AB002303.1 - 4 551
AF434817.1 - 2 303
BC030808.1 - 3 413 4701661
BX537424.1 - 3 435
BX956000.1 - 3 482
CR933621.1 - 4 486
DR005686.1 - 4 478

View Full Product Details