Gene Transcripts

Gene Symbol
Ccin
Entrez Gene ID
442829
Gene Name
calicin
Gene Aliases
4933417A14Rik
Chromosome Location
Chr. 4: 43983504 - 43985533 on Build GRCm38
UniGene ID
Mm.348215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002787.2 NP_001002787.1 1 809
GenBank mRNA AK030197.1 - 1 812
BC150949.1 - 1 796 9087860
BC150952.1 - 1 796 9087863

View Full Product Details