Gene Transcripts
- Gene Symbol
- ZFYVE16
- Entrez Gene ID
- 9765
- Gene Name
- zinc finger FYVE-type containing 16
- Gene Aliases
- PPP1R69
- Chromosome Location
- Chr. 5: 80407615 - 80479869 on Build GRCh38
- UniGene ID
- Hs.482660
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001105251.2 |
NP_001098721.1
|
10, 11 |
3685 |
|
NM_001284236.1 |
NP_001271165.1
|
10, 11 |
3611 |
|
NM_014733.4 |
NP_055548.3
|
9, 10 |
3557 |
|
XM_005248632.4 |
XP_005248689.1
|
9, 10 |
3683 |
|
XM_011543753.2 |
XP_011542055.1
|
10, 11 |
3582 |
|
XM_017010091.1 |
XP_016865580.1
|
11, 12 |
3743 |
|
XM_017010092.1 |
XP_016865581.1
|
10, 11 |
3729 |
|
XM_017010093.1 |
XP_016865582.1
|
9, 10 |
2063 |
|
XM_017010094.1 |
XP_016865583.1
|
8, 9 |
2008 |
|
XM_017010095.1 |
XP_016865584.1
|
11, 12 |
3637 |
|
XM_017010096.1 |
XP_016865585.1
|
6, 7 |
1085 |
|
GenBank mRNA |
AB002303.1 |
- |
10, 11 |
3625 |
|
AF434817.1 |
- |
8, 9 |
3377 |
|
AK127003.1 |
- |
5, 6 |
775 |
|
AL833087.2 |
- |
7, 8 |
2604 |
|
BX537424.1 |
- |
9, 10 |
3509 |
|
CR933621.1 |
- |
10, 11 |
3560 |
|
View Full Product Details