Gene Transcripts

Gene Symbol
Fam98c
Entrez Gene ID
292764
Gene Name
family with sequence similarity 98, member C
Gene Aliases
RGD1303117
Chromosome Location
Chr. 1: 88095240 - 88098816 on Build Rnor_6.0
UniGene ID
Rn.12517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198584.1 NP_001185513.1 8 1224
XM_006228717.2 XP_006228779.1 9 1156
GenBank mRNA BC079080.1 - 7 1262

View Full Product Details