Gene Transcripts

Gene Symbol
Samm50
Entrez Gene ID
300111
Gene Name
SAMM50 sorting and assembly machinery component
Gene Aliases
Chromosome Location
Chr. 7: 125058966 - 125082506 on Build Rnor_6.0
UniGene ID
Rn.12611
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004241.1 NP_001004241.1 6 570
GenBank mRNA BC079302.1 - 6 570

View Full Product Details