Gene Transcripts
- Gene Symbol
- ZYG11A
- Entrez Gene ID
- 440590
- Gene Name
- zyg-11 family member A, cell cycle regulator
- Gene Aliases
- ZYG11
- Chromosome Location
- Chr. 1: 52842511 - 52894575 on Build GRCh38
- UniGene ID
- Hs.658458
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY271826.1 |
- |
14 |
3378 |
|
View Full Product Details