Gene Transcripts

Gene Symbol
Clec4a4
Entrez Gene ID
474145
Gene Name
C-type lectin domain family 4, member a4
Gene Aliases
Dcir2
Chromosome Location
Chr. 6: 122990367 - 123024421 on Build GRCm38
UniGene ID
Mm.263483
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005860.2 NP_001005860.1 1 40
GenBank mRNA AY397673.1 - 1 40
BC104370.1 - 1 40 40049888
BC104371.2 - 1 40

View Full Product Details