Gene Transcripts

Gene Symbol
Foxn3
Entrez Gene ID
71375
Gene Name
forkhead box N3
Gene Aliases
5430426H20Rik, AA589593, AW556347, Ches1, Ches1l, HTLFL1
Chromosome Location
Chr. 12: 99190081 - 99563625 on Build GRCm38
UniGene ID
Mm.341972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183186.2 NP_899009.2 6 2376
GenBank mRNA AK046748.1 - 6 2651
AK077921.1 - 1 1239
BC099971.1 - 6 2454 30008715

View Full Product Details