Gene Transcripts

Gene Symbol
KRT39
Entrez Gene ID
390792
Gene Name
keratin 39
Gene Aliases
CK-39, K39, KA35
Chromosome Location
Chr. 17: 40958417 - 40966892 on Build GRCh38
UniGene ID
Hs.28467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213656.3 NP_998821.3 1 248
GenBank mRNA AJ786657.1 - 1 248
BC157887.1 - 1 242 8862708
BC171907.1 - 1 242

View Full Product Details