Gene Transcripts
- Gene Symbol
- Inca1
- Entrez Gene ID
- 103844
- Gene Name
- inhibitor of CDK, cyclin A1 interacting protein 1
- Gene Aliases
- AI842396
- Chromosome Location
- Chr. 11: 70688361 - 70700206 on Build GRCm38
- UniGene ID
- Mm.101581
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252482.1 |
NP_001239411.1
|
3, 4 |
408 |
|
NM_001252483.1 |
NP_001239412.1
|
3, 4 |
369 |
|
NM_001252484.1 |
NP_001239413.1
|
3, 4 |
299 |
|
NM_001252485.1 |
NP_001239414.1
|
4, 5 |
528 |
|
NM_213729.1 |
NP_998894.1
|
3, 4 |
437 |
|
GenBank mRNA |
AK139002.1 |
- |
3, 4 |
297 |
|
AY601909.1 |
- |
3, 4 |
437 |
|
AY601910.1 |
- |
3, 4 |
408 |
|
AY601911.1 |
- |
3, 4 |
369 |
|
AY601912.1 |
- |
3, 4 |
299 |
|
BC096427.1 |
- |
4, 5 |
482 |
4160694 |
BC115572.1 |
- |
3, 4 |
268 |
40102153 |
BC115573.1 |
- |
3, 4 |
268 |
40102154 |
View Full Product Details