Gene Transcripts

Gene Symbol
CBY1
Entrez Gene ID
25776
Gene Name
chibby family member 1, beta catenin antagonist
Gene Aliases
C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1
Chromosome Location
Chr. 22: 38656653 - 38673850 on Build GRCh38
UniGene ID
Hs.334911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002880.1 NP_001002880.2 4 429
NM_015373.3 NP_056188.1 3 289
GenBank mRNA AB111855.1 - 2 167
AF331041.1 - 3 225
AK307265.1 - 3 304
AK311928.1 - 3 302
AL050345.1 - 3 268
AL136686.1 - 3 272
BC016139.1 - 3 267 3922707
CD702742.1 - 3 327
CR456410.1 - 2 181

View Full Product Details