Gene Transcripts
- Gene Symbol
- VPS39
- Entrez Gene ID
- 23339
- Gene Name
- VPS39, HOPS complex subunit
- Gene Aliases
- TLP, VAM6, hVam6p
- Chromosome Location
- Chr. 15: 42158701 - 42208331 on Build GRCh38
- UniGene ID
- Hs.88025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301138.1 |
NP_001288067.1
|
14, 15 |
1578 |
|
NM_015289.3 |
NP_056104.2
|
13, 14 |
1545 |
|
XM_011521403.2 |
XP_011519705.1
|
14, 15 |
1547 |
|
XM_011521404.2 |
XP_011519706.1
|
13, 14 |
1550 |
|
XM_017022036.1 |
XP_016877525.1
|
14, 15 |
1546 |
|
XM_017022037.1 |
XP_016877526.1
|
13, 14 |
1550 |
|
XM_017022038.1 |
XP_016877527.1
|
13, 14 |
1733 |
|
XM_017022039.1 |
XP_016877528.1
|
13, 14 |
1544 |
|
GenBank mRNA |
AB018313.2 |
- |
13, 14 |
1496 |
|
AF280814.1 |
- |
14, 15 |
1405 |
|
AF281052.1 |
- |
13, 14 |
1515 |
|
BC015817.2 |
- |
9, 10 |
1105 |
|
BC068559.1 |
- |
13, 14 |
1514 |
5295166 |
BX537404.1 |
- |
13, 14 |
1524 |
|
View Full Product Details