Gene Transcripts

Gene Symbol
VGLL1
Entrez Gene ID
51442
Gene Name
vestigial like family member 1
Gene Aliases
TDU, VGL1
Chromosome Location
Chr. X: 136532152 - 136556807 on Build GRCh38
UniGene ID
Hs.496843
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016267.3 NP_057351.1 5 966
GenBank mRNA AF137387.1 - 5 862
BC000045.1 - 5 966 3504527
BC003362.1 - 5 864 2900293

View Full Product Details