Gene Transcripts

Gene Symbol
Actrt1
Entrez Gene ID
302796
Gene Name
actin-related protein T1
Gene Aliases
Chromosome Location
Chr. X: 133227699 - 133229052 on Build Rnor_6.0
UniGene ID
Rn.110052
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013961.1 NP_001013983.1 1 830
GenBank mRNA BC083679.1 - 1 830

View Full Product Details