Gene Transcripts
- Gene Symbol
- GDAP2
- Entrez Gene ID
- 54834
- Gene Name
- ganglioside induced differentiation associated protein 2
- Gene Aliases
- MACROD3
- Chromosome Location
- Chr. 1: 117860227 - 117930042 on Build GRCh38
- UniGene ID
- Hs.310809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135589.2 |
NP_001129061.1
|
12, 13 |
1609 |
|
NM_017686.3 |
NP_060156.1
|
12, 13 |
1609 |
|
XM_005270959.2 |
XP_005271016.1
|
12, 13 |
1609 |
|
XM_011541621.2 |
XP_011539923.1
|
12, 13 |
1609 |
|
XM_011541622.1 |
XP_011539924.1
|
12, 13 |
1497 |
|
XM_011541623.2 |
XP_011539925.1
|
12, 13 |
1609 |
|
XM_011541624.2 |
XP_011539926.1
|
13, 14 |
2052 |
|
XM_017001531.1 |
XP_016857020.1
|
13, 14 |
1631 |
|
XM_017001532.1 |
XP_016857021.1
|
13, 14 |
2052 |
|
GenBank mRNA |
AK000149.1 |
- |
12, 13 |
1512 |
|
BC013132.1 |
- |
12, 13 |
1492 |
3448678 |
View Full Product Details