Gene Transcripts

Gene Symbol
Shc1
Entrez Gene ID
85385
Gene Name
SHC (Src homology 2 domain containing) transforming protein 1
Gene Aliases
P66shc
Chromosome Location
Chr. 2: 188745503 - 188757066 on Build Rnor_6.0
UniGene ID
Rn.138818
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164060.1 NP_001157532.1 12 2230
NM_053517.2 NP_445969.2 13 1797
GenBank mRNA BC088298.1 - 13 1797

View Full Product Details