Gene Transcripts

Gene Symbol
OR4K2
Entrez Gene ID
390431
Gene Name
olfactory receptor family 4 subfamily K member 2
Gene Aliases
OR14-15
Chromosome Location
Chr. 14: 19876268 - 19877212 on Build GRCh38
UniGene ID
Hs.551345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005501.1 NP_001005501.1 1 346
GenBank mRNA BC136941.1 - 1 382
BC136942.1 - 1 382 9020934

View Full Product Details