Gene Transcripts

Gene Symbol
Cryz
Entrez Gene ID
362061
Gene Name
crystallin, zeta (quinone reductase)
Gene Aliases
Chromosome Location
Chr. 2: 260884318 - 260911827 on Build Rnor_6.0
UniGene ID
Rn.6746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012183.2 NP_001012183.1 9 1417
XM_006233520.2 XP_006233582.1 10 1653
GenBank mRNA BC078927.1 - 9 1419

View Full Product Details