Gene Transcripts

Gene Symbol
Ccndbp1
Entrez Gene ID
362201
Gene Name
cyclin D-type binding-protein 1
Gene Aliases
Chromosome Location
Chr. 3: 112950970 - 112961620 on Build Rnor_6.0
UniGene ID
Rn.162679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013204.1 NP_001013222.1 11 1032
XM_006234859.1 XP_006234921.1 10 1080
GenBank mRNA BC091256.1 - 11 1032
FQ233751.1 - 11 1073

View Full Product Details