Gene Transcripts

Gene Symbol
Fam71b
Entrez Gene ID
432552
Gene Name
family with sequence similarity 71, member B
Gene Aliases
Gari-l3, OTTMUSG00000005491
Chromosome Location
Chr. 11: 46404728 - 46407985 on Build GRCm38
UniGene ID
Mm.443244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013783.2 NP_001013805.1 2 1885
GenBank mRNA AB473727.1 - 3 1789
BC150988.1 - 3 1817 9087901

View Full Product Details